Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008008.4 | NP_032034.1 | 2 | 621 | ||
GenBank mRNA | AK033934.1 | - | 1 | 900 | ||
AK037172.1 | - | 2 | 623 | |||
AK086639.1 | - | 2 | 508 | |||
BC052847.1 | - | 2 | 504 | 30054875 | ||
Z22703.1 | - | 1 | 158 |