Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008008.4 | NP_032034.1 | 4 | 909 | ||
GenBank mRNA | AK033934.1 | - | 3 | 1189 | ||
AK037172.1 | - | 4 | 911 | |||
AK086639.1 | - | 4 | 796 | |||
BC052847.1 | - | 4 | 792 | 30054875 | ||
Z22703.1 | - | 3 | 446 |