Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008011.2 | NP_032037.2 | 3 | 484 | ||
GenBank mRNA | AK084850.1 | - | 3 | 484 | ||
AK085723.1 | - | 3 | 483 | |||
AY493377.2 | - | 2 | 370 | |||
BC033313.1 | - | 3 | 671 | 5132831 | ||
DQ388428.1 | - | 2 | 370 | |||
X59927.1 | - | 3 | 426 |