Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301412.1 | NP_001288341.1 | 3, 4 | 571 | ||
NM_008017.4 | NP_032043.3 | 3, 4 | 495 | |||
GenBank mRNA | AJ534939.1 | - | 2, 3 | 326 | ||
AK013109.1 | - | 3, 4 | 537 | |||
AK017305.1 | - | 3, 4 | 541 | |||
AK017747.1 | - | 3, 4 | 474 | |||
AK019977.1 | - | 3, 4 | 543 | |||
AK028233.1 | - | 3, 4 | 537 | |||
AK145341.1 | - | 3, 4 | 525 | |||
AK152090.1 | - | 3, 4 | 524 | |||
AK161661.1 | - | 3, 4 | 465 | |||
BC011064.1 | - | 3, 4 | 433 | |||
BC021847.1 | - | 3, 4 | 583 | |||
BC094380.1 | - | 3, 4 | 467 | 30543190 | ||
CJ049961.1 | - | 3, 4 | 496 |