Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002777.1 | - | 5 | 760 | ||
AK010693.1 | - | 5 | 785 | |||
AK145400.1 | - | 5 | 778 | |||
AK145986.1 | - | 4 | 811 | |||
AK151047.1 | - | 3 | 572 | |||
AK154751.1 | - | 5 | 733 | |||
AK168333.1 | - | 5 | 769 | |||
AK169186.1 | - | 5 | 692 | |||
AK169242.1 | - | 5 | 776 | |||
BC004671.1 | - | 5 | 736 | 3498682 | ||
X60203.1 | - | 5 | 751 |