Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008022.2 | NP_032048.1 | 1 | 1597 | ||
GenBank mRNA | BC138432.1 | - | 1 | 1485 | 8861452 | |
BC138434.1 | - | 1 | 1485 | 8861454 |