Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040403.1 | NP_001035493.1 | 10 | 1297 | ||
NM_001284227.1 | NP_001271156.1 | 11 | 1381 | |||
NM_001284228.1 | NP_001271157.1 | 11 | 1244 | |||
NM_008028.2 | NP_032054.1 | 11 | 1388 | |||
GenBank mRNA | AK004589.1 | - | 10 | 1235 | ||
AK030199.1 | - | 10 | 1260 | |||
AK149530.1 | - | 11 | 1319 | |||
AK170557.1 | - | 10 | 1298 | |||
BC070423.1 | - | 11 | 1346 | 6403816 | ||
U07890.1 | - | 11 | 1258 |