Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040403.1 | NP_001035493.1 | 11 | 2243 | ||
NM_001284227.1 | NP_001271156.1 | 12 | 2327 | |||
NM_001284228.1 | NP_001271157.1 | 12 | 2190 | |||
NM_008028.2 | NP_032054.1 | 12 | 2334 | |||
GenBank mRNA | AK004589.1 | - | 11 | 2181 | ||
AK030199.1 | - | 11 | 2206 | |||
AK149530.1 | - | 12 | 2265 | |||
AK170557.1 | - | 11 | 2244 | |||
AK179634.1 | - | 1 | 123 | |||
AK181207.1 | - | 1 | 128 | |||
AK182347.1 | - | 1 | 125 | |||
AK183299.1 | - | 1 | 120 | |||
AK193346.1 | - | 1 | 64 | |||
AK198617.1 | - | 1 | 327 | |||
AK205894.1 | - | 1 | 92 | |||
BC070423.1 | - | 12 | 2292 | 6403816 | ||
U07890.1 | - | 12 | 2204 |