Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008030.1 | NP_032056.1 | 5 | 566 | ||
GenBank mRNA | AK050123.1 | - | 5 | 595 | ||
AK149440.1 | - | 5 | 593 | |||
U87147.1 | - | 5 | 566 |