Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290424.1 | NP_001277353.1 | 5 | 576 | ||
NM_008031.3 | NP_032057.2 | 5 | 576 | |||
GenBank mRNA | AF461114.1 | - | 5 | 300 | ||
AK046605.1 | - | 5 | 536 | |||
AK053211.1 | - | 2 | 318 | |||
AK053701.1 | - | 5 | 536 | |||
AK053829.1 | - | 5 | 578 | |||
AK080948.1 | - | 5 | 536 | |||
AK140343.1 | - | 5 | 538 | |||
BC079671.1 | - | 5 | 562 | 30532682 | ||
L23971.1 | - | 5 | 423 |