Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290424.1 | NP_001277353.1 | 17 | 2059 | ||
NM_008031.3 | NP_032057.2 | 17 | 2134 | |||
GenBank mRNA | BC079671.1 | - | 17 | 2120 | 30532682 | |
L23971.1 | - | 17 | 1981 |