Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008037.4 | NP_032063.2 | 4 | 3466 | ||
GenBank mRNA | AK137197.1 | - | 1 | 992 | ||
AK162508.1 | - | 1 | 992 | |||
BC065131.1 | - | 4 | 3404 | 30536144 |