Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008044.2 | NP_032070.1 | 5 | 875 | ||
GenBank mRNA | BC058533.1 | - | 5 | 900 | ||
U95736.1 | - | 5 | 875 |