Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122892.1 | NP_001116364.1 | 8 | 1244 | ||
NM_008054.2 | NP_032080.2 | 9 | 1314 | |||
GenBank mRNA | AK171646.1 | - | 9 | 1316 | ||
BC010554.1 | - | 8 | 925 | |||
BC032149.1 | - | 8 | 959 | 3602546 | ||
BC092217.1 | - | 8 | 1240 | 6403711 | ||
M27266.1 | - | 10 | 1026 | |||
U70324.1 | - | 9 | 1027 |