Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122892.1 | NP_001116364.1 | 13 | 3277 | ||
NM_001122893.1 | NP_001116365.1 | 14 | 3356 | |||
NM_008054.2 | NP_032080.2 | 14 | 3347 | |||
GenBank mRNA | AK040199.1 | - | 1 | 1028 | ||
AK156584.1 | - | 14 | 3357 | |||
U70324.1 | - | 14 | 3070 |