Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162494.1 | NP_001155966.1 | 4 | 1020 | ||
NM_008056.3 | NP_032082.3 | 4 | 1024 | |||
GenBank mRNA | AK088399.1 | - | 4 | 889 | ||
BC026150.1 | - | 4 | 1013 | 3983985 | ||
U43319.1 | - | 3 | 850 |
Set Membership: |
G-protein coupled receptor |