Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293621.1 | NP_001280550.1 | 12 | 1528 | ||
NM_008060.2 | NP_032086.1 | 13 | 1594 | |||
GenBank mRNA | AK028655.1 | - | 12 | 1498 | ||
AK030722.1 | - | 12 | 1497 | |||
AK047454.1 | - | 7 | 849 | |||
AK081915.1 | - | 12 | 1490 | |||
AK122201.1 | - | 7 | 1724 | |||
AK149686.1 | - | 13 | 1551 | |||
AK166150.1 | - | 12 | 1467 | |||
AK178842.1 | - | 1 | 83 | |||
AK181935.1 | - | 1 | 83 | |||
BC020386.1 | - | 5 | 744 | |||
BC035423.1 | - | 3 | 344 | |||
BC094437.1 | - | 12 | 1478 | 30682580 | ||
BC117888.1 | - | 12 | 1471 | 40096413 | ||
BC117889.1 | - | 12 | 1474 | 40096414 | ||
BC128069.1 | - | 13 | 1538 | 40096428 | ||
U92793.1 | - | 13 | 1540 |