Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008067.4 | NP_032093.3 | 10 | 1733 | ||
GenBank mRNA | AK039144.1 | - | 10 | 1674 | ||
AK134760.1 | - | 10 | 1642 | |||
BC119317.1 | - | 10 | 1531 | 8734066 | ||
BC120686.1 | - | 10 | 1531 | 40129609 | ||
CD354278.1 | - | 2 | 459 | |||
M86568.1 | - | 10 | 1635 |