Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005117.2 | NP_005108.1 | 3 | 822 | ||
GenBank mRNA | AB018122.1 | - | 3 | 359 | ||
AF110400.1 | - | 3 | 822 | |||
AY358302.1 | - | 3 | 822 | |||
BC017664.1 | - | 3 | 490 | 3849343 | ||
BT006729.1 | - | 3 | 359 |