Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008075.2 | NP_032101.3 | 6, 7 | 762 | ||
GenBank mRNA | AF024620.1 | - | 6, 7 | 762 | ||
BI736525.1 | - | 6, 7 | 796 |