Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008078.2 | NP_032104.2 | 16 | 2490 | ||
GenBank mRNA | D42051.1 | - | 16 | 2507 |