Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008078.2 | NP_032104.2 | 4 | 826 | ||
GenBank mRNA | AF326549.1 | - | 4 | 490 | ||
AF326550.1 | - | 4 | 490 | |||
AK136834.1 | - | 4 | 526 | |||
AK141473.1 | - | 4 | 571 | |||
BC018380.1 | - | 4 | 541 | 4482097 | ||
D42051.1 | - | 4 | 842 | |||
KC134211.1 | - | 4 | 490 | |||
L16980.1 | - | 4 | 562 |