Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005121.2 | NP_005112.2 | 3 | 486 | ||
XM_011525551.2 | XP_011523853.1 | 3 | 1187 | |||
XM_011525552.2 | XP_011523854.1 | 3 | 1187 | |||
XM_011525553.2 | XP_011523855.1 | 1 | 47 | |||
GenBank mRNA | AF117754.1 | - | 4 | 486 | ||
BC140891.1 | - | 3 | 460 | 9021759 | ||
BQ709424.1 | - | 2 | 242 |