Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008098.4 | NP_032124.1 | 4 | 2173 | ||
GenBank mRNA | AK049398.1 | - | 4 | 2073 | ||
AK083888.1 | - | 4 | 2087 | |||
AK145755.1 | - | 4 | 2078 | |||
AK146305.1 | - | 4 | 2120 | |||
AK146473.1 | - | 4 | 2076 | |||
AK150750.1 | - | 4 | 2094 | |||
AK151106.1 | - | 4 | 2041 | |||
AK151147.1 | - | 4 | 2041 | |||
AK151485.1 | - | 4 | 2098 | |||
AK151564.1 | - | 4 | 2041 | |||
AK151640.1 | - | 4 | 2096 | |||
AK151687.1 | - | 4 | 2069 | |||
AK152272.1 | - | 4 | 2069 | |||
AK152317.1 | - | 4 | 2058 | |||
BC043084.1 | - | 4 | 2070 | 6417343 | ||
BC054811.1 | - | 4 | 2086 | 6839739 | ||
D78188.1 | - | 4 | 2081 |