Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcsam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159297.1 | NP_001152769.1 | 6 | 1060 | ||
NM_008099.4 | NP_032125.3 | 5 | 994 | |||
GenBank mRNA | AK036553.1 | - | 5 | 1012 | ||
BC099585.1 | - | 5 | 958 | 30289367 | ||
U13263.1 | - | 5 | 995 |