Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008109.2 | NP_032135.2 | 2 | 956 | ||
GenBank mRNA | AB259648.1 | - | 2 | 647 | ||
BC034546.1 | - | 2 | 968 | 4190036 | ||
U08337.1 | - | 2 | 968 |