Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008109.2 | NP_032135.2 | 2 | 2260 | ||
GenBank mRNA | AK041168.1 | - | 1 | 1214 | ||
BC034546.1 | - | 2 | 2276 | 4190036 | ||
CJ326163.1 | - | 1 | 449 | |||
U08337.1 | - | 2 | 2276 |