Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008112.4 | NP_032138.3 | 4 | 517 | ||
GenBank mRNA | AK007871.1 | - | 4 | 460 | ||
AK013104.1 | - | 4 | 462 | |||
AK078144.1 | - | 460 | ||||
AK078514.1 | - | 4 | 464 | |||
AK079690.1 | - | 3 | 358 | |||
AK083063.1 | - | 4 | 459 | |||
AK090083.1 | - | 4 | 465 | |||
AK137761.1 | - | 4 | 464 | |||
AK143633.1 | - | 4 | 462 | |||
AK143681.1 | - | 4 | 496 | |||
AK149958.1 | - | 4 | 470 | |||
AK150356.1 | - | 4 | 452 | |||
AK150655.1 | - | 4 | 487 | |||
AK152250.1 | - | 4 | 451 | |||
AK153381.1 | - | 4 | 430 | |||
AK159734.1 | - | 4 | 462 | |||
AK166317.1 | - | 4 | 462 | |||
AK167659.1 | - | 4 | 474 | |||
AK169299.1 | - | 4 | 428 | |||
AK186127.1 | - | 2 | 109 | |||
BC024971.1 | - | 4 | 438 | |||
BC053381.1 | - | 2617 | ||||
BC055341.1 | - | 4 | 443 | 6808475 | ||
L36314.1 | - | 4 | 323 |