Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008112.4 | NP_032138.3 | 11 | 2086 | ||
GenBank mRNA | AF095728.1 | - | 1 | 533 | ||
AK007871.1 | - | 11 | 2029 | |||
AK078144.1 | - | 2029 | ||||
AK079690.1 | - | 10 | 1927 | |||
AK083063.1 | - | 11 | 2028 | |||
AK137761.1 | - | 11 | 2033 | |||
AK143633.1 | - | 11 | 2031 | |||
AK143681.1 | - | 11 | 2065 | |||
AK149958.1 | - | 11 | 2039 | |||
AK150356.1 | - | 11 | 2021 | |||
AK150655.1 | - | 11 | 2056 | |||
AK152250.1 | - | 11 | 2019 | |||
AK159734.1 | - | 11 | 2031 | |||
AK166317.1 | - | 11 | 2043 | |||
AK167659.1 | - | 11 | 2043 | |||
AK169299.1 | - | 11 | 1997 | |||
BC024971.1 | - | 11 | 2007 | |||
BC053381.1 | - | 4185 | ||||
BC055341.1 | - | 11 | 2012 | 6808475 |