Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005131.2 | NP_005122.2 | 2 | 117 | ||
XM_011525772.2 | XP_011524074.1 | 2 | 117 | |||
XM_011525773.1 | XP_011524075.1 | 2 | 105 | |||
XM_017026104.1 | XP_016881593.1 | 2 | 117 | |||
GenBank mRNA | AK225067.1 | - | 2 | 93 | ||
AK314755.1 | - | 2 | 83 | |||
AY573302.1 | - | 2 | 77 | |||
AY573303.1 | - | 2 | 77 | |||
BC010381.1 | - | 2 | 108 | 4046908 | ||
DA902294.1 | - | 2 | 117 | |||
L36529.1 | - | 2 | 91 |