Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271628.1 | NP_001258557.1 | 3 | 442 | ||
NM_008121.3 | NP_032147.1 | 2 | 385 | |||
GenBank mRNA | AK038780.1 | - | 1114 | |||
AK137069.1 | - | 2 | 346 | |||
BC053054.1 | - | 2 | 371 | 6401526 |