Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159382.1 | NP_001152854.1 | 3 | 1180 | ||
NM_001159383.1 | NP_001152855.1 | 3 | 1173 | |||
NM_008122.3 | NP_032148.1 | 3 | 1729 | |||
GenBank mRNA | AK052353.1 | - | 1 | 534 | ||
BC023153.1 | - | 5 | 984 | |||
BC050840.1 | - | 3 | 1180 | 6330453 | ||
BC071230.1 | - | 3 | 1173 | 30443940 | ||
X63100.1 | - | 3 | 1350 |