Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glycam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289587.1 | NP_001276516.1 | 4 | 569 | ||
NM_008134.3 | NP_032160.1 | 4 | 531 | |||
GenBank mRNA | AK021358.1 | - | 4 | 529 | ||
BC031931.1 | - | 2 | 1983 | |||
BC145548.1 | - | 4 | 498 | 9054112 | ||
BC145832.1 | - | 4 | 504 | 40131135 | ||
BC145858.1 | - | 4 | 504 | 40131161 | ||
M93428.1 | - | 4 | 503 |