Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008138.4 | NP_032164.2 | 9 | 1669 | ||
GenBank mRNA | AK159222.1 | - | 9 | 1673 | ||
AK160186.1 | - | 9 | 1666 | |||
AK167388.1 | - | 9 | 1671 | |||
BC006695.1 | - | 2 | 640 | |||
BC037130.1 | - | 5 | 993 | |||
BC065159.1 | - | 9 | 1656 | 5717305 |