Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008160.6 | NP_032186.2 | 2 | 553 | ||
GenBank mRNA | AK002245.1 | - | 2 | 358 | ||
AK010999.1 | - | 2 | 358 | |||
AK011019.1 | - | 2 | 358 | |||
AK028171.1 | - | 2 | 358 | |||
AK150548.1 | - | 2 | 359 | |||
AK154833.1 | - | 2 | 358 | |||
AK160388.1 | - | 2 | 359 | |||
BC086649.1 | - | 2 | 341 | 6819448 | ||
CK628430.1 | - | 2 | 553 |