Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 4 | 598 | ||
NM_008162.3 | NP_032188.3 | 4 | 544 | |||
GenBank mRNA | AB072797.1 | - | 4 | 580 | ||
AF045768.1 | - | 4 | 462 | |||
AF045769.1 | - | 4 | 353 | |||
AF274027.1 | - | 4 | 574 | |||
AK006441.1 | - | 4 | 439 | |||
AK145770.1 | - | 4 | 464 | |||
AK160389.1 | - | 4 | 371 | |||
AK160509.1 | - | 4 | 347 | |||
AK168027.1 | - | 4 | 371 | |||
BC083137.1 | - | 3 | 331 | |||
BC106147.1 | - | 4 | 391 | 4936866 | ||
CA466677.1 | - | 4 | 525 | |||
D87896.1 | - | 4 | 544 |