Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 7 | 881 | ||
NM_008162.3 | NP_032188.3 | 7 | 827 | |||
GenBank mRNA | AA510431.1 | - | 4 | 251 | ||
AB072797.1 | - | 7 | 863 | |||
AF045768.1 | - | 7 | 745 | |||
AF045769.1 | - | 7 | 636 | |||
AK006441.1 | - | 7 | 722 | |||
AK145770.1 | - | 7 | 748 | |||
AK160389.1 | - | 7 | 654 | |||
AK160509.1 | - | 7 | 630 | |||
AK167881.1 | - | 8 | 686 | |||
AK168027.1 | - | 7 | 654 | |||
BC083137.1 | - | 6 | 614 | |||
BC106147.1 | - | 7 | 674 | 4936866 | ||
D87896.1 | - | 7 | 827 |