Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 5 | 771 | ||
GenBank mRNA | AK077750.1 | - | 4 | 591 | ||
AK079087.1 | - | 4 | 599 | |||
AK148148.1 | - | 5 | 736 | |||
AK153557.1 | - | 4 | 704 | |||
AK155800.1 | - | 5 | 726 | |||
AK166142.1 | - | 5 | 725 | |||
AK170845.1 | - | 4 | 562 | |||
AK172002.1 | - | 5 | 747 | |||
AK181485.1 | - | 1 | 64 | |||
AK212651.1 | - | 1 | 96 | |||
AK216513.1 | - | 1 | 96 | |||
AK217799.1 | - | 1 | 96 | |||
BC052377.1 | - | 5 | 771 | 5696248 | ||
BC085254.1 | - | 5 | 607 | 5343274 | ||
D85748.1 | - | 3 | 395 | |||
U07617.1 | - | 4 | 520 |