Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113325.2 | NP_001106796.1 | 16 | 3671 | ||
NM_001252403.1 | NP_001239332.1 | 16 | 3298 | |||
NM_008165.4 | NP_032191.2 | 16 | 3671 | |||
GenBank mRNA | AK135107.1 | - | 16 | 3300 | ||
BC056397.1 | - | 16 | 3336 | 6842391 | ||
BC060702.1 | - | 16 | 3254 |