Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008174.2 | NP_032200.2 | 4 | 1155 | ||
GenBank mRNA | AK020963.1 | - | 4 | 1140 | ||
AK038395.1 | - | 4 | 1208 | |||
AY673682.1 | - | 3 | 783 | |||
BC051384.1 | - | 4 | 1155 | 6466902 | ||
BC137596.1 | - | 3 | 808 | 8860615 | ||
BC144753.1 | - | 3 | 808 | 9053301 | ||
BC144754.1 | - | 3 | 808 | 9053302 | ||
U17252.1 | - | 3 | 853 |
Set Membership: |
G-protein coupled receptor |