Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008187.2 | NP_032213.2 | 5 | 797 | ||
GenBank mRNA | AK011217.1 | - | 5 | 741 | ||
AK088713.1 | - | 5 | 701 | |||
AK172603.1 | - | 4 | 1607 | |||
BC092397.1 | - | 3 | 3213 | |||
BC119023.1 | - | 5 | 693 | 8733772 | ||
BC119025.1 | - | 5 | 693 | 8733774 | ||
BC144770.1 | - | 4 | 613 | |||
CB195345.1 | - | 2 | 219 | |||
Z54179.1 | - | 5 | 720 |