Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 1065 | ||
GenBank mRNA | AK051213.1 | - | 1 | 828 | ||
AK077473.1 | - | 1 | 742 | |||
AK090029.1 | - | 1 | 832 | |||
AK145675.1 | - | 1 | 717 | |||
AK146077.1 | - | 1 | 828 | |||
AK153990.1 | - | 1 | 829 | |||
AK155312.1 | - | 1 | 760 | |||
BC003830.1 | - | 1 | 814 | 3600538 | ||
BC011493.1 | - | 1 | 812 | 4166167 | ||
BC110361.1 | - | 1 | 817 | 5343233 | ||
X13171.1 | - | 1 | 795 |