Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142706.1 | NP_001136178.1 | 3, 4 | 891 | ||
NM_008198.2 | NP_032224.2 | 3, 4 | 891 | |||
GenBank mRNA | AK098094.1 | - | 3, 4 | 551 | ||
AK149531.1 | - | 3, 4 | 524 | |||
AK149573.1 | - | 3, 4 | 550 | |||
AK151246.1 | - | 3, 4 | 625 | |||
BC005451.1 | - | 3, 4 | 525 | 3591669 | ||
M57890.1 | - | 3, 4 | 594 |