Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142706.1 | NP_001136178.1 | 7, 8 | 1427 | ||
NM_008198.2 | NP_032224.2 | 7, 8 | 1427 | |||
GenBank mRNA | AK098094.1 | - | 7, 8 | 1087 | ||
AK149531.1 | - | 7, 8 | 1060 | |||
AK149573.1 | - | 7, 8 | 1086 | |||
AK151220.1 | - | 1, 2 | 46 | |||
AK151246.1 | - | 7, 8 | 1161 | |||
AK151618.1 | - | 1, 2 | 46 | |||
AK151974.1 | - | 1, 2 | 46 | |||
BC005451.1 | - | 7, 8 | 1061 | 3591669 | ||
K01496.1 | - | 2, 3 | 167 | |||
K01497.1 | - | 1, 2 | 26 | |||
M57890.1 | - | 7, 8 | 1130 |