Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008209.4 | NP_032235.1 | 6 | 2309 | ||
GenBank mRNA | AF010448.1 | - | 6 | 2220 | ||
AF010451.1 | - | 3 | 3007 | |||
AF010453.1 | - | 2038 | ||||
AK137171.1 | - | 6 | 2150 | |||
BC026137.1 | - | 6 | 2337 | 4017492 |