Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 4 | 641 | ||
NM_005168.4 | NP_005159.1 | 5 | 548 | |||
GenBank mRNA | AF498969.1 | - | 4 | 363 | ||
AK299731.1 | - | 4 | 348 | |||
AK309442.1 | - | 3 | 222 | |||
AK313224.1 | - | 4 | 615 | |||
AL702945.1 | - | 4 | 614 | |||
BC012513.1 | - | 5 | 503 | 4453777 | ||
BQ440721.1 | - | 4 | 617 | |||
BT006769.1 | - | 4 | 363 | |||
CR542070.1 | - | 4 | 318 | |||
S82240.1 | - | 4 | 342 | |||
X95282.1 | - | 4 | 363 | |||
X97758.1 | - | 4 | 585 |