Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008221.4 | NP_032247.1 | 2 | 179 | ||
GenBank mRNA | AK011619.1 | - | 2 | 180 | ||
AK012501.1 | - | 2 | 181 | |||
AK012897.1 | - | 2 | 180 | |||
AK077938.1 | - | 270 | ||||
BC057014.1 | - | 2 | 156 | 6817810 | ||
M26897.1 | - | 2 | 141 | |||
V00857.1 | - | 2 | 61 |