Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008225.2 | NP_032251.2 | 9 | 756 | ||
GenBank mRNA | AK088723.1 | - | 9 | 738 | ||
BC007469.1 | - | 9 | 726 | 3593456 | ||
D42120.1 | - | 9 | 687 | |||
X84797.1 | - | 9 | 724 |