Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008229.2 | NP_032255.2 | 3 | 425 | ||
GenBank mRNA | AB221663.1 | - | 3 | 217 | ||
AK051753.1 | - | 3 | 400 | |||
BC138517.1 | - | 3 | 397 | 8861537 | ||
U31758.1 | - | 3 | 385 |