Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008229.2 | NP_032255.2 | 14 | 1928 | ||
GenBank mRNA | AK141649.1 | - | 4 | 545 | ||
U31758.1 | - | 14 | 1889 |